Anti-C2CD2L Mouse Polyclonal Antibody
Catalog # ABNOH00009854-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:C2CD2-like
- Antigen symbol:C2CD2L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9854
- Antigen synonyms:TMEM24|DLNB23
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: C2CD2L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human