Anti-TCL1B Mouse Monoclonal Antibody [clone: 2A12]
Catalog # ABNOH00009623-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:T-cell leukemia/lymphoma 1B
- Antigen symbol:TCL1B
- Clonality:Monoclonal
- Clone:2A12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9623
- Antigen synonyms:TML1|SYN-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: TCL1B
Clonality: Monoclonal
Clone: 2A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human