Anti-BAG5 Mouse Monoclonal Antibody [clone: 2E2]
Catalog # ABNOH00009529-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:BCL2 Associated Athanogene 5
- Antigen symbol:BAG5
- Clonality:Monoclonal
- Clone:2E2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9529
- Antigen synonyms:BAG family molecular chaperone regulator 5| BCL2 associated athanogene 5|BAG5_HUMAN.|BAG 5|KIAA0873|BAG-5
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: BAG5
Clonality: Monoclonal
Clone: 2E2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human