Anti-VPS4B Mouse Polyclonal Antibody
ABNOH00009525-A01
: Abnova
- Antibody type:Primary
- Antigen name:Vacuolar Protein Sorting 4B
- Antigen symbol:VPS4B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9525
- Antigen synonyms:MIG1|VPS4-2|SKD1B|SKD1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: VPS4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human