Anti-VAPB Mouse Monoclonal Antibody [clone: 1A2]
Catalog # ABNOH00009217-M10
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:VAMP (vesicle-associated membrane Protein)-associated protein B
- Antigen symbol:VAPB
- Clonality:Monoclonal
- Clone:1A2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9217
- Antigen synonyms:ALS8|VAP-B|VAMP-B
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: VAPB
Clonality: Monoclonal
Clone: 1A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human