Anti-SLMAP Mouse Monoclonal Antibody [clone: 2A7]
Catalog # ABNOH00007871-M08
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Sarcolemma Associated Protein
- Clonality:Monoclonal
- Clone:2A7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:7871
- Antigen synonyms:SLAP
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: SLMAP
Clonality: Monoclonal
Clone: 2A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human