Anti-TYK2 Mouse Monoclonal Antibody [clone: 6H1]
ABNOH00007297-M03
: Abnova
- Antibody type:Primary
- Antigen name:Tyrosine Kinase 2
- Antigen symbol:TYK2
- Clonality:Monoclonal
- Clone:6H1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:7297
- Antigen synonyms:IMD35|TYK2|JTK1|non-receptor tyrosine kinase 2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: TYK2
Clonality: Monoclonal
Clone: 6H1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human