Order Entry
Ireland
ContactUsLinkComponent
 
Anti-TEAD4 Mouse Monoclonal Antibody [clone: 5H3]
  ABNOH00007004-M01A
 :  Abnova
undefined
Anti-TEAD4 Mouse Monoclonal Antibody [clone: 5H3]
  ABNOH00007004-M01A
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    TEA domain family member 4
  • Antigen symbol:
    TEAD4
  • Clonality:
    Monoclonal
  • Clone:
    5H3
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    7004
  • Antigen synonyms:
    TCF13L1|TEFR-1|EFTR-2|TEF-3|RTEF1|TEF3|hRTEF-1B
  • Storage buffer:
    In ascites fluid
  • Sequence:
    ARGPGRPAVSGFWQGALPGQAGTSHDVKPFSQQTYAVQPPLPLPGFESPAGPAPSPSAPPAPPWQGRSVASSKLWMLEFSAFLEQQQDPDTYNKHLFVHIGQSSPSYSDP
  • Storage temperature:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Shipping temperature:
    Ice
  • Immunogen:
    TEAD4 (NP_003204, 151 a.a. ~ 260 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
  • Size:
    200 µl
  • Pk:
    200 µl

 

 

Type: Primary
Antigen: TEAD4
Clonality: Monoclonal
Clone: 5H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human