Anti-AKR1D1 Rabbit Polyclonal Antibody
ABNOH00006718-D01P
: Abnova
- Antibody type:Primary
- Antigen name:Aldo-keto reductase family 1 member D1
- Antigen symbol:AKR1D1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:6718
- Antigen synonyms:3o5bred|CBAS2|SRD5B1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: AKR1D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human