Anti-SRD5A1 Rabbit Polyclonal Antibody
Catalog # ABNOH00006715-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Steroid 5 alpha-reductase 1
- Antigen symbol:SRD5A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:6715
- Antigen synonyms:SR type 1|S5A1_HUMAN.|Steroid 5-alpha-reductase 1|S5AR 1|3-oxo-5-alpha-steroid 4-dehydrogenase 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: SRD5A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human