Order Entry
Ireland
ContactUsLinkComponent
Anti-RBMS1 Mouse Monoclonal Antibody [clone: 3F2-2G9]
Anti-RBMS1 Mouse Monoclonal Antibody [clone: 3F2-2G9]
Catalog # ABNOH00005937-M01
Supplier:  Abnova
undefined
Anti-RBMS1 Mouse Monoclonal Antibody [clone: 3F2-2G9]
Catalog # ABNOH00005937-M01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    RNA binding motif, single stranded interacting protein 1
  • Antigen symbol:
    RBMS1
  • Clonality:
    Monoclonal
  • Clone:
    3F2-2G9
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    5937
  • Antigen synonyms:
    C2orf12|MSSP-1|SCR2|YC1|MSSP-3|MSSP-2|HCC-4|MSSP
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: RBMS1
Clonality: Monoclonal
Clone: 3F2-2G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human