Anti-MAPK9 Mouse Monoclonal Antibody [clone: 1D6]
Catalog # ABNOH00005601-M05
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase 9
- Antigen symbol:MAPK9
- Clonality:Monoclonal
- Clone:1D6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5601
- Antigen synonyms:SAPK1a|JNK-55|JNK2A|PRKM9|p54aSAPK|SAPK|p54a|JNK2|JNK2BETA|JNK2B|JNK2ALPHA
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: MAPK9
Clonality: Monoclonal
Clone: 1D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat