Anti-PRKAG1 Rabbit Polyclonal Antibody
Catalog # ABNOH00005571-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Protein Kinase, AMP-activated, Non-catalytic Subunit Gamma 1
- Antigen symbol:PRKAG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5571
- Antigen synonyms:5'-AMP-activated protein kinase subunit gamma-1|AMPKG3|AMPK gamma3|PRKAG3|AMPK- gamma3|5'-AMP-activated protein kinase subunit gamma-2|AMPK- gamma1|AMPK subunit gamma-1|AMPKg|AMPK subunit gamma-3|H91620p|PRKAG2|AMPK- gamma2|AMPK subunit gamma-2|PRKAG1|AMPK gamma2|AMPK gamma1|5'-AMP-activated protein kinase subunit gamma-3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PRKAG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human