Anti-PRF1 Mouse Monoclonal Antibody [clone: 3B4]
Catalog # ABNOH00005551-M04
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Perforin 1
- Antigen symbol:PRF1
- Clonality:Monoclonal
- Clone:3B4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5551
- Antigen synonyms:P1|FLH2|PFP|HPLH2|PFN1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PRF1
Clonality: Monoclonal
Clone: 3B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human