Anti-POLH Rabbit Polyclonal Antibody
Catalog # ABNOH00005429-D01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Polymerase (DNA) Eta
- Antigen symbol:POLH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:5429
- Antigen synonyms:RAD30A|RAD30|XP-V|XPV|XPV/DNA polymerase eta
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: POLH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human