Anti-PFDN2 Mouse Monoclonal Antibody [clone: 2C1]
Catalog # ABNOH00005202-M07
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Prefoldin Subunit 2
- Antigen symbol:PFDN2
- Clonality:Monoclonal
- Clone:2C1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5202
- Antigen synonyms:PFD2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PFDN2
Clonality: Monoclonal
Clone: 2C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human