Anti-PDHA2 Mouse Polyclonal Antibody
ABNOH00005161-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Pyruvate dehydrogenase (lipoamide) alpha 2
- Antigen symbol:PDHA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5161
- Antigen synonyms:PDHAL
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: PDHA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human