Anti-NRAS Mouse Polyclonal Antibody
ABNOH00004893-A01
: Abnova
- Antibody type:Primary
- Antigen name:Neuroblastoma RAS viral (v-ras) Oncogene homolog
- Antigen symbol:NRAS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:4893
- Antigen synonyms:NRAS1|Neuroblastoma RAS|N-RAS|NS6|RASN|GTPase Nras|RAS|NRAS|ALPS4|Neuroblastoma RAS viral (v-ras) oncogene homolog|Transforming protein N-Ras
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: NRAS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human