Anti-NDUFS5 Rabbit Polyclonal Antibody
ABNOH00004725-D01P
: Abnova
- Antibody type:Primary
- Antigen name:NADH Dehydrogenase (Ubiquinone) Fe-S Protein 5, 15Kda (Nadh-Coenzyme Q Reductase)
- Antigen symbol:NDUFS5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:4725
- Antigen synonyms:CI-15k|CI15K
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: NDUFS5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human