Anti-NAGA Mouse Polyclonal Antibody
ABNOH00004668-B01P
: Abnova
- Antibody type:Primary
- Antigen name:N-acetylgalactosaminidase, alpha
- Antigen symbol:NAGA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:4668
- Antigen synonyms:GALB|D22S674
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: NAGA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human