Anti-LOXL1 Rabbit Polyclonal Antibody
ABNOH00004016-D01P
: Abnova
- Antibody type:Primary
- Antigen name:lysyl oxidase-like 1
- Antigen symbol:LOXL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:4016
- Antigen synonyms:LOL|LOXL
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: LOXL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human