Anti-GPT Mouse Monoclonal Antibody [clone: 1F10]
Catalog # ABNOH00002875-M02A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Glutamic-pyruvate Transaminase
- Antigen symbol:GPT
- Clonality:Monoclonal
- Clone:1F10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2875
- Antigen synonyms:Glutamic--pyruvic transaminase 1|AAT1|GPT1|GPT|Alanine aminotransferase 1|Glutamic--alanine transaminase 1 |Glutamate pyruvate transaminase 1|ALT1|GPT 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: GPT
Clonality: Monoclonal
Clone: 1F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human