Anti-GNG4 Mouse Monoclonal Antibody [clone: 1D9-G11]
Catalog # ABNOH00002786-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Guanine nucleotide binding protein (G protein), gamma 4
- Antigen symbol:GNG4
- Clonality:Monoclonal
- Clone:1D9-G11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2786
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: GNG4
Clonality: Monoclonal
Clone: 1D9-G11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human