Anti-GNA11 Rabbit Polyclonal Antibody
Catalog # ABNOH00002767-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:G Protein Subunit Alpha 11
- Antigen symbol:GNA11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2767
- Antigen synonyms:GNA-11|GNA11|GNA11_HUMAN|Guanine nucleotide-binding protein Gy subunit alpha|HYPOC2|HHC2|Guanine nucleotide-binding protein subunit alpha-11.|FBH2|FHH2|FBH|G-protein subunit alpha-11|G alpha-11
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: GNA11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human