Anti-GDI1 Mouse Monoclonal Antibody [clone: 1H3]
Catalog # ABNOH00002664-M08
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:GDP dissociation inhibitor 1
- Antigen symbol:GDI1
- Clonality:Monoclonal
- Clone:1H3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2664
- Antigen synonyms:MRX48|GDIL|RABGD1A|RABGDIA|MRX41|FLJ41411|XX-FW81657B9.1|XAP-4|OPHN2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: GDI1
Clonality: Monoclonal
Clone: 1H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human