Anti-GATA2 Mouse Monoclonal Antibody [clone: 1G5]
Catalog # ABNOH00002624-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:GATA binding Protein 2
- Antigen symbol:GATA2
- Clonality:Monoclonal
- Clone:1G5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2624
- Antigen synonyms:MONOMAC|GATA-binding protein 2|NFE1B|DCML|endothelial transcription factor GATA-2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: GATA2
Clonality: Monoclonal
Clone: 1G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human