Anti-FABP7 Mouse Polyclonal Antibody
ABNOH00002173-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Fatty Acid Binding Protein 7
- Antigen symbol:FABP7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:2173
- Antigen synonyms:BLBP|LTR2-FABP7|FABPB|B-FABP|MRG
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: FABP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human