Anti-EEF1G Mouse Monoclonal Antibody [clone: S1]
Catalog # ABNOH00001937-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Elongation factor 1-gamma
- Antigen symbol:EEF1G
- Clonality:Monoclonal
- Clone:S1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:1937
- Antigen synonyms:GIG35|EF1G|EF-1-GAMMA
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: EEF1G
Clonality: Monoclonal
Clone: S1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human