Order Entry
Ireland
ContactUsLinkComponent
Anti-PHC2 Mouse Monoclonal Antibody [clone: 1F4]
Anti-PHC2 Mouse Monoclonal Antibody [clone: 1F4]
Catalog # ABNOH00001912-M01
Supplier:  Abnova
undefined
Anti-PHC2 Mouse Monoclonal Antibody [clone: 1F4]
Catalog # ABNOH00001912-M01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Polyhomeotic Homolog 2
  • Antigen symbol:
    PHC2
  • Clonality:
    Monoclonal
  • Clone:
    1F4
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Gene ID:
    1912
  • Antigen synonyms:
    EDR2|PH2|HPH2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: PHC2
Clonality: Monoclonal
Clone: 1F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human