Anti-SLC26A2 Mouse Monoclonal Antibody [clone: 3F6]
ABNOH00001836-M04
: Abnova
- Antibody type:Primary
- Antigen name:Solute Carrier Family 26 Member A2
- Antigen symbol:Solute carrier family 26, member 2
- Clonality:Monoclonal
- Clone:3F6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:1836
- Antigen synonyms:DTDST|D5S1708|DTD|EDM4|MST153|MSTP157
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: SLC26A2
Clonality: Monoclonal
Clone: 3F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human