Order Entry
Ireland
ContactUsLinkComponent
Anti-AMPD2 Mouse Monoclonal Antibody [clone: 6A8]
Anti-AMPD2 Mouse Monoclonal Antibody [clone: 6A8]
Catalog # ABNOH00000271-M09A
Supplier:  Abnova
undefined
Anti-AMPD2 Mouse Monoclonal Antibody [clone: 6A8]
Catalog # ABNOH00000271-M09A
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    AMP deaminase 2
  • Antigen symbol:
    AMPD2
  • Clonality:
    Monoclonal
  • Clone:
    6A8
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    271
  • Antigen synonyms:
    PCH9|SPG63
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    200 µl
  • Pk:
    200 µl

Specifications

About this item

Type: Primary
Antigen: AMPD2
Clonality: Monoclonal
Clone: 6A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human