Anti-TYRO3 Mouse Monoclonal Antibody [clone: OTI2G2]
ORIGTA500420
: OriGene
- Antibody type:Primary
- Antigen name:TYRO3 Protein Tyrosine Kinase
- Antigen symbol:TYRO3
- Clonality:Monoclonal
- Clone:OTI2G2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:96.7 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150; Immunoprecipitation: 2?g/500?l
Type: Primary
Antigen: TYRO3 (TYRO3 protein tyrosine kinase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human