Anti-LGR5 Mouse Antibody
ORIGTA400031
: OriGene
- Antibody type:Primary
- Antigen name:leucine-rich repeat containing G Protein-coupled receptor 5
- Antigen symbol:LGR5
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Type: Primary
Antigen: LGR5 (leucine-rich repeat containing G protein-coupled receptor 5)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: