Anti-MUC5AC Mouse Monoclonal Antibody [clone: 1-13M1]
Catalog # ORIGTA317044
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:mucin 5AC, oligomeric mucus/gel-forming
- Antigen symbol:MUC5AC
- Clonality:Monoclonal
- Clone:1-13M1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS, 0,05% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Pk:50 µG
Specifications
About this item
Recommended Dilutions: Immunochemistry: 1:100;
Type: Primary
Antigen: MUC5AC (mucin 5AC, oligomeric mucus/gel-forming)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human