Anti-PLA2G4C Rabbit Polyclonal Antibody
Catalog # ORIGTA314779
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Phospholipase A2, group IVC (cytosolic, calcium-independent)
- Antigen symbol:PLA2G4C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Form:Liquid
- Storage buffer:Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:1000
Type: Primary
Antigen: PLA2G4C (phospholipase A2, group IVC (cytosolic, calcium-independent))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human