Anti-OAZ1 Rabbit Polyclonal Antibody
ORIGTA314297
: OriGene
- Antibody type:Primary
- Antigen name:ornithine decarboxylase antizyme 1
- Antigen symbol:OAZ1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:none
- Form:Liquid
- Storage buffer:Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:40000
Type: Primary
Antigen: OAZ1 (ornithine decarboxylase antizyme 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat