Anti-ADAMTS3 Rabbit Polyclonal Antibody
Catalog # ORIGTA310093
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:ADAM metallopeptidase with thrombospondin type 1 motif, 3
- Antigen symbol:ADAMTS3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS, pH 7,4 with 0,05% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: ADAMTS3 (ADAM metallopeptidase with thrombospondin type 1 motif, 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human