Order Entry
Ireland
ContactUsLinkComponent
 
Anti-NAT2 Rabbit Polyclonal Antibody
  ORIGTA308634
 :  OriGene
Anti-NAT2 Rabbit Polyclonal Antibody
  ORIGTA308634
 :  OriGene

 

  • Antibody type:
    Primary
  • Antigen name:
    N-acetyltransferase 2 (arylamine N-acetyltransferase)
  • Antigen symbol:
    NAT2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Amino acid number:
    1 to 198
  • Storage buffer:
    0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7). 0,01% Thimerosal was added as a preservative.
  • Molecular weight:
    34 kDa
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by antigen affinity chromatography
  • Pk:
    100 µl

 

 

Recommended Dilutions: Immunochemistry1:100-1:1000; Western Blot:1:500-1:3000

Type: Primary
Antigen: NAT2 (N-acetyltransferase 2 (arylamine N-acetyltransferase))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human