Order Entry
Ireland
ContactUsLinkComponent
 
Anti-PSMF1 Rabbit Polyclonal Antibody
Catalog # ORIGTA308605
Supplier:  OriGene
Anti-PSMF1 Rabbit Polyclonal Antibody
Catalog # ORIGTA308605
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Proteasome (prosome, macropain) Inhibitor Subunit 1
  • Antigen symbol:
    PI31
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Amino acid number:
    209 to 271
  • Storage buffer:
    0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7). 0,01% Thimerosal was added as a preservative.
  • Molecular weight:
    30 kDa
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by antigen affinity chromatography
  • Pk:
    100 µl

Specifications

About this item

Recommended Dilutions: Immunochemistry/Immunofluorescence:1:100-1:1000; Western Blot:1:500-1:3000

Type: Primary
Antigen: PSMF1 (proteasome (prosome, macropain) inhibitor subunit 1 (PI31))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human