Anti-CXCR4 Rabbit Polyclonal Antibody
Catalog # ORIGTA305936
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:chemokine (C-X-C motif) receptor 4
- Antigen symbol:CXCR4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Amino acid number:182 to 196
- Storage buffer:PBS containing 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by DEAE chromatography
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500-1:1000
Type: Primary
Antigen: CXCR4 (chemokine (C-X-C motif) receptor 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse