Order Entry
Ireland
ContactUsLinkComponent
Anti-APC Rabbit Monoclonal Antibody [clone: EP701Y]
Anti-APC Rabbit Monoclonal Antibody [clone: EP701Y]
Catalog # ORIGTA303394
Supplier:  OriGene
Anti-APC Rabbit Monoclonal Antibody [clone: EP701Y]
Catalog # ORIGTA303394
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    adenomatous polyposis coli
  • Antigen symbol:
    APC
  • Clonality:
    Monoclonal
  • Clone:
    EP701Y
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    near N-terminus
  • Cross adsorption:
    none
  • Format:
    Tissue culture supernatant
  • Form:
    Liquid
  • Storage buffer:
    PBS 49%,Sodium azide 0,01%,Glycerol 50%,BSA 0,05%
  • Molecular weight:
    160 kDa
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µG

Specifications

About this item

Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:50-100; Immunoprecipitation: 1:50

Type: Primary
Antigen: APC (adenomatous polyposis coli)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat