Anti-MHC II (I-A/I-E) Rat Monoclonal Antibody (violetFluor® 450) [clone: M5/115.15.2]
Supplier: Tonbo Biosciences
Certificates
About this item
The M5/114.15.2 antibody reacts with mouse MHC Class II alloantigens I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek, as well as being cross-reactive with mouse cells of H-2p and H-2r haplotype. MHC Class II is widely expressed by mouse immune cells bearing these alloantigens, including T and B cells, monocytes, macrophages and dendritic cells. The antibody does not react with the following alloantigens: I-Af, I-Ak, I-As, or NOD H-2g7.
Type: Primary
Antigen: MHC II (I-A/I-E) (MHC II (I-A/I-E))
Clonality: monoclonal
Clone: M5/115.15.2
Conjugation: violetFluor® 450
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity: Mouse
Specifications
- Catalog No:
- 75-5321-U100
- 75-5321-U025
- Antigen symbol:
- MHC2
- MHC2
- Antigen name:
- MHC Class 2
- MHC Class 2
- Antigen synonyms:
- MHC II|IA/IE|IA|IE|I-A/E
- MHC II|IA/IE|IA|IE|I-A/E
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- VioletFluor® 450
- VioletFluor® 450
- Clone:
- M5/115.15.2
- M5/115.15.2
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 14961/14969
- 14961/14969
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Cross adsorption:
- none
- none
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together