Order Entry
Ireland
ContactUsLinkComponent
Anti-CD27 Mouse Monoclonal Antibody [clone: O323]
Anti-CD27 Mouse Monoclonal Antibody [clone: O323]
Catalog # 70-0279-U100
Supplier:  Tonbo Biosciences
undefined
Anti-CD27 Mouse Monoclonal Antibody [clone: O323]
Catalog # 70-0279-U100
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD27 molecule
  • Antigen symbol:
    CD27
  • Clonality:
    Monoclonal
  • Clone:
    O323
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Baboon,
    Cynomolgus monkey,
    Macaque,
    Human
  • Environmentally Preferable:
  • Cross adsorption:
    none
  • Gene ID:
    939
  • Antigen synonyms:
    Tp55|S152|T14|TNFRSF7|CD27|S152. LPFS2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µG

Specifications

About this item

The O323 antibody reacts with human CD27 (TNFRSF7), a cell surface homodimer of 55 kDa subunits, which provides co-stimulatory signaling in support of the T cell (TCR) and B cell (BCR) receptors. By comparison with CD28, whose TCR co-stimulatory signal can trigger cell proliferation, CD27 signaling appears to promote cell survival and differentiation to effector / memory stages. Also in contrast with CD28, the CD27 receptor may be shed following interaction with its ligand CD70, which is typically expressed on activated dendritic cells, T cells and B cells. With respect to B cells, CD27 is considered to be a phenotypic marker for memory B cells. CD27 has been included within a group of phenotypic markers for identifying human B regulatory cells (Bregs), a cell type proposed to regulate CD4+ T cell proliferation and Foxp3 / CTLA-4 expression in Treg cells.

The O323 antibody may be used for analysis of CD27 expression on peripheral T cells, and is frequently used in combination with antibodies for IgD and IgM to distinguish naïve vs. memory B cell populations. For identification of Breg cells, this antibody has been used in combination with antibodies for CD25, CD1d, IL-10 and TGF-beta (Kessel et al. 2012. Autoimm. Rev. 11(9): 670-677). The antibody is also reported for cross-reactivity with Baboon, Cynomolgus and Rhesus CD27.

Type: Primary
Antigen: CD27 (CD27 molecule)
Clonality: monoclonal
Clone: O323
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: