About this item
The J43.1 antibody is specific for mouse CD279, also known as programmed death-1 (PD-1), a 55 kDa glycoprotein which can co-regulate T cell antigen receptor signaling and therefore modulate T cell activation. PD-1 exists in a monomeric form that is expressed by CD4- CD8- thymocytes, where it participates in the processes of clonal selection, elimination of autoreactive lymphocytes, and development of tolerance. PD-1 expression is also inducible upon activation of mature T cells, where it has been proposed to interact with the co-stimulatory receptor CD80 to limit T cell activation. Two ligands for PD-1, known as PD-L1 (B7-H1) and PD-L2 (B7-DC) are differentially expressed on T and B cells, monocytes, macrophages, NK cells or dendritic cells. PD-1 is a member of a family of receptors including CD28 and CTLA-4 (CD152), which interact with 'B7' ligands to provide a balance of co-stimulatory/co-inhibitory signaling important in T cell activation, tolerance, and autoimmunity.
The J43.1 antibody may be used as a marker for PD-1 expression, and is commonly used for analysis of receptor-ligand interaction and function(s) in vitro and in vivo. Please choose the appropriate format for each application.
Type: Primary
Antigen: PDCD1 (programmed cell death 1)
Clonality: monoclonal
Clone: J43
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 50-9985-U025
- 50-9985-U100
- Antigen symbol:
- PD1
- PD1
- Antigen name:
- Programmed cell death protein 1
- Programmed cell death protein 1
- Antigen synonyms:
- PD-1|hPD-1|hPD-l|programmed cell death protein 1|CD279|protein PD-1|PD1|SLEB2
- PD-1|hPD-1|hPD-l|programmed cell death protein 1|CD279|protein PD-1|PD1|SLEB2
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- J43
- J43
- Reactivity:
- Mouse
- Mouse
- Host:
- Armenian hamster
- Armenian hamster
- Gene ID:
- 18566
- 18566
- Isotype:
- IgG
- IgG
- Flow cytometry:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Cross adsorption:
- none
- none
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
- Tested applications:
- Flow Cytometry
- Flow Cytometry