Order Entry
Ireland
ContactUsLinkComponent
Anti-CD86 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: PO3.1]
Anti-CD86 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: PO3.1]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD86 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: PO3.1]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 50-0861-U025
  • 50-0861-U100
  • Antigen symbol:
  • CD86
  • CD86
  • Antigen name:
  • CD86 molecule
  • CD86 molecule
  • Antigen synonyms:
  • CLS1|B70|B7.2|Ly-58|Cd28l2|ETC-1|MB7|TS/A-2|B7-2|B7|MB7-2|Ly58
  • CLS1|B70|B7.2|Ly-58|Cd28l2|ETC-1|MB7|TS/A-2|B7-2|B7|MB7-2|Ly58
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • PO3.1
  • PO3.1
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 12524
  • 12524
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together