Order Entry
Ireland
ContactUsLinkComponent
Anti-CD274 Rat Monoclonal Antibody [clone: 10F.9G2]
Anti-CD274 Rat Monoclonal Antibody [clone: 10F.9G2]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD274 Rat Monoclonal Antibody [clone: 10F.9G2]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 40-1243-U500
  • 40-1243-U100
  • Antigen symbol:
  • CD274
  • CD274
  • Antigen name:
  • CD274 molecule
  • CD274 molecule
  • Antigen synonyms:
  • PDL1|PD-L1|B7H1|B7-H|PDCD1L1|PDCD1LG1
  • PDL1|PD-L1|B7H1|B7-H|PDCD1L1|PDCD1LG1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 10F.9G2
  • 10F.9G2
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 60533
  • 60533
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together