Order Entry
Ireland
ContactUsLinkComponent
Anti-ICAM1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 15.2]
Anti-ICAM1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 15.2]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-ICAM1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 15.2]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 20-0549-T025
  • 20-0549-T100
  • Antigen symbol:
  • ICAM1
  • ICAM1
  • Antigen name:
  • Intercellular Adhesion molecule 1
  • Intercellular Adhesion molecule 1
  • Antigen synonyms:
  • ICAM-1|major group rhinovirus receptor|human rhinovirus receptor|BB2|cell surface glycoprotein P3.58|intercellular adhesion molecule 1|P3.58
  • ICAM-1|major group rhinovirus receptor|human rhinovirus receptor|BB2|cell surface glycoprotein P3.58|intercellular adhesion molecule 1|P3.58
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • 15.2
  • 15.2
  • Reactivity:
  • Pig, Human
  • Pig, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 3383
  • 3383
  • Isotype:
  • IgG1
  • IgG1
  • Flow cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice
  • Tested applications:
  • Flow cytometry
  • Flow cytometry

Specifications

Related Information

Frequently Bought Together