Anti-CAPN3 Rabbit Polyclonal Antibody
ABGEAP9817B
: Abgent
- Antibody type:Primary
- Antigen name:Calpain 3
- Antigen symbol:CAPN3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-Terminal
- Gene ID:3621
- Antigen synonyms:CANP3|p94|LGMD2A|LGMD2|nCL-1|CANPL3
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:SPAERLVAEADEGGPC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This ING1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human ING1.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: CAPN3 (calpain 3, (p94))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human