Anti-SERPINA1 Rabbit Polyclonal Antibody
Catalog # ABGEAP9431D
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1
- Antigen symbol:SERPINA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Gene ID:6605
- Antigen synonyms:alpha1AT|PRO2275|AAT|PI|A1A|PI1|A1AT
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:EEQAANKGEEKKDDC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This SMARCE1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 307-335 amino acids from the C-terminal region of human SMARCE1.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: SERPINA1 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human