Anti-TREF1 Rabbit Polyclonal Antibody
Catalog # ABGEAP9356B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Transcriptional Regulating Factor 1
- Antigen symbol:TRERF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Gene ID:126129
- Antigen synonyms:HSA277276|dJ139D8.5|RAPA|TREP132|TReP-132|BCAR2
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:NFVRAMEDKEKTDPQC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,4 mg/ml
- Shipping temperature:4 °C
- Immunogen:This CPT1C antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 596-625 amino acids from the C-terminal region of human CPT1C.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: TREF1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human